COM448682569 Combi-Blocks, Inc.

Boc-3-methoxy-d-phenylalanine

  • MDL number MFCD07371964
  • Cas Number : 261380-37-4
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7301
Availability details for
Quantity
  • Availability
  • Add to Cart