COM448682621 Combi-Blocks, Inc.

Ethyl 3,5-dimethylpyrazole-4-carboxylate, HCl

  • MDL number MFCD16495875
  • Cas Number : 1305320-57-3
  • Molecular Weight : 204.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7373
Availability details for
Quantity
  • Availability
  • Add to Cart