COM448682661 Combi-Blocks, Inc.

Cis-1-tert-butyl 3-ethyl 4-aminopyrrolidine-1,3-dicarboxylate, HCl

  • MDL number MFCD15530331
  • Cas Number : 1233501-65-9
  • Molecular Weight : 294.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7445
Availability details for
Quantity
  • Availability
  • Add to Cart