COM448682663 Combi-Blocks, Inc.

Cis-1-tert-butyl 4-methyl 3-methylpiperidine-1,4-dicarboxylate

  • MDL number MFCD20261477
  • Cas Number : 1334499-78-3
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7447
Availability details for
Quantity
  • Availability
  • Add to Cart