COM448682685 Combi-Blocks, Inc.

cis-Bicyclo[3.1.0]hexan-3-ol

  • MDL number MFCD20921684
  • Cas Number : 694-43-9
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7476
Availability details for
Quantity
  • Availability
  • Add to Cart