COM448682687 Combi-Blocks, Inc.

Cis-ethyl 3-((tert-butoxycarbonyl)amino)tetrahydro-2h-pyran-4-carboxylate

  • MDL number MFCD21607477
  • Cas Number : 1363404-85-6
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7479
Availability details for
Quantity
  • Availability
  • Add to Cart