COM448682689 Combi-Blocks, Inc.

Cis-ethyl 3-methylpiperidine-4-carboxylate hcl

  • MDL number MFCD20261475
  • Cas Number : 1159822-87-3
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7482
Availability details for
Quantity
  • Availability
  • Add to Cart