COM448682693 Combi-Blocks, Inc.

2-Amino-5-methoxycarbonyl benzoic acid

  • MDL number MFCD08690064
  • Cas Number : 63746-25-8
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7487
Availability details for
Quantity
  • Availability
  • Add to Cart