COM448682723 Combi-Blocks, Inc.

Cyclopropyl(piperidin-4-yl)methanone hcl

  • MDL number MFCD21607363
  • Cas Number : 1363405-16-6
  • Molecular Weight : 189.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7535
Availability details for
Quantity
  • Availability
  • Add to Cart