COM448682735 Combi-Blocks, Inc.

1-Benzyl-3-(chloromethyl)-1h-indazole

  • MDL number MFCD20134218
  • Cas Number : 131427-22-0
  • Molecular Weight : 256.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7552

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7552
Availability details for
Quantity
  • Availability
  • Add to Cart