COM448682751 Combi-Blocks, Inc.

D-1-N-Cbz-nipecotamide

  • MDL number MFCD09025344
  • Cas Number : 1050446-94-0
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7575
Availability details for
Quantity
  • Availability
  • Add to Cart