COM448682765 Combi-Blocks, Inc.

4-Fluoro-2-methanesulfonamidobenzoic acid

  • MDL number MFCD09734189
  • Cas Number : 170107-84-3
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7604

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7604
Availability details for
Quantity
  • Availability
  • Add to Cart