COM448682768 Combi-Blocks, Inc.

D-Cysteine, HCl

  • MDL number MFCD00012632
  • Cas Number : 32443-99-5
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7607

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7607
Availability details for
Quantity
  • Availability
  • Add to Cart