COM448682835 Combi-Blocks, Inc.

2-Amino-4-(1h-imidazol-1-yl)benzoic acid

  • MDL number MFCD11656801
  • Cas Number : 1141669-47-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7701
Availability details for
Quantity
  • Availability
  • Add to Cart