COM448682849 Combi-Blocks, Inc.

(R)-N-Boc-5-hydroxy-trp-ome

  • MDL number MFCD12198870
  • Cas Number : 1234880-33-1
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7723
Availability details for
Quantity
  • Availability
  • Add to Cart