COM448682864 Combi-Blocks, Inc.

Dimethyl 2-methyladipate

  • MDL number MFCD00025877
  • Cas Number : 19780-94-0
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7741
Availability details for
Quantity
  • Availability
  • Add to Cart