COM448682865 Combi-Blocks, Inc.

Dimethyl 2-phenoxysuccinate

  • MDL number MFCD09998265
  • Cas Number : 96019-08-8
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7742
Availability details for
Quantity
  • Availability
  • Add to Cart