COM448682883 Combi-Blocks, Inc.

6-Chloroimidazo[1,2-a]pyridine-5-carbaldehyde

  • MDL number MFCD22887424
  • Cas Number : 1414864-02-0
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7772
Availability details for
Quantity
  • Availability
  • Add to Cart