COM448682909 Combi-Blocks, Inc.

Ethyl (r)-(3-n-boc-amino-piperidin-2-yl)-acetate

  • MDL number MFCD07369951
  • Cas Number : 1219132-56-5
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7814
Availability details for
Quantity
  • Availability
  • Add to Cart