COM448682927 Combi-Blocks, Inc.

Ethyl 1,1-dioxo-isothiazolidine-3-carboxylate

  • MDL number MFCD09878821
  • Cas Number : 1253789-57-9
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7839
Availability details for
Quantity
  • Availability
  • Add to Cart