COM448682962 Combi-Blocks, Inc.

Ethyl 1-nitro-4-oxo-4h-quinolizine-3-carboxylate

  • MDL number MFCD09788597
  • Cas Number : 1556-30-5
  • Molecular Weight : 262.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7887
Availability details for
Quantity
  • Availability
  • Add to Cart