COM448682963 Combi-Blocks, Inc.

Ethyl 1-phenylimidazole-4-carboxylate

  • MDL number MFCD11041365
  • Cas Number : 197079-08-6
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7889
Availability details for
Quantity
  • Availability
  • Add to Cart