COM448682969 Combi-Blocks, Inc.

Ethyl 2-((s)-3-amino-2-oxopiperidin-1-yl)acetate hcl

  • MDL number MFCD11973826
  • Cas Number : 937057-79-9
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7896
Availability details for
Quantity
  • Availability
  • Add to Cart