COM448682978 Combi-Blocks, Inc.

Methyl 5-(aminomethyl)-2-methoxybenzoate, HCl

  • MDL number MFCD07285352
  • Cas Number : 1049782-92-4
  • Molecular Weight : 231.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7913
Availability details for
Quantity
  • Availability
  • Add to Cart