COM448682981 Combi-Blocks, Inc.

Ethyl 2-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenylamino)acetate

  • MDL number MFCD11867865
  • Cas Number : 1092460-58-6
  • Molecular Weight : 345.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7918
Availability details for
Quantity
  • Availability
  • Add to Cart