COM448682986 Combi-Blocks, Inc.

Ethyl 2-(4-chlorophenyl)-2-hydroxyacetate

  • MDL number MFCD00068980
  • Cas Number : 13511-29-0
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7930
Availability details for
Quantity
  • Availability
  • Add to Cart