COM448682989 Combi-Blocks, Inc.

Methyl 4-benzylmorpholine-3-carboxylate

  • MDL number MFCD09756515
  • Cas Number : 212650-44-7
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7936

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7936
Availability details for
Quantity
  • Availability
  • Add to Cart