COM448682999 Combi-Blocks, Inc.

Ethyl 2-(pyrazin-2-yl)acetate

  • MDL number MFCD10698624
  • Cas Number : 1060815-23-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7952
Availability details for
Quantity
  • Availability
  • Add to Cart