COM448683015 Combi-Blocks, Inc.

1-(5,6-Dimethyl-thieno[2,3-d]pyrimidin-4-yl)-piperidine-4-carboxylic acid

  • MDL number MFCD05840360
  • Cas Number : 842971-60-2
  • Molecular Weight : 291.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7978

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7978
Availability details for
Quantity
  • Availability
  • Add to Cart