COM448683020 Combi-Blocks, Inc.

Ethyl 2-amino-5-fluoro-2,3-dihydro-1h-indene-2-carboxylate hcl

  • MDL number MFCD18711559
  • Cas Number : 1272758-28-7
  • Molecular Weight : 259.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7988
Availability details for
Quantity
  • Availability
  • Add to Cart