COM448683029 Combi-Blocks, Inc.

Ethyl 2-bromo-4-(trifluoromethyl)oxazole-5-carboxylate

  • MDL number MFCD17926293
  • Cas Number : 1227934-69-1
  • Molecular Weight : 288.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-7999
Availability details for
Quantity
  • Availability
  • Add to Cart