COM448683050 Combi-Blocks, Inc.

Ethyl 2-guanidino-4-methylthiazole-5-carboxylate

  • MDL number MFCD08131777
  • Cas Number : 131184-96-8
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8033
Availability details for
Quantity
  • Availability
  • Add to Cart