COM448683078 Combi-Blocks, Inc.

Ethyl 3-(1-Boc-piperidin-4-yl)-D,L-beta-alaninate

  • MDL number MFCD07369851
  • Cas Number : 886362-37-4
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8078
Availability details for
Quantity
  • Availability
  • Add to Cart