COM448683086 Combi-Blocks, Inc.

Ethyl 3-(2-methoxyphenyl)-4-nitrobutanoate

  • MDL number MFCD11501918
  • Cas Number : 290366-76-6
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8088
Availability details for
Quantity
  • Availability
  • Add to Cart