COM448683101 Combi-Blocks, Inc.

Ethyl 3-(4-chloro-phenyl)-2-methylaminomethyl-propionate

  • MDL number MFCD08061166
  • Cas Number : 886364-67-6
  • Molecular Weight : 255.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8108
Availability details for
Quantity
  • Availability
  • Add to Cart