COM448683108 Combi-Blocks, Inc.

Ethyl 3-(6-chloropyrazin-2-yloxy)benzoate

  • MDL number MFCD11501089
  • Cas Number : 1086397-56-9
  • Molecular Weight : 278.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8118

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8118
Availability details for
Quantity
  • Availability
  • Add to Cart