COM448683161 Combi-Blocks, Inc.

Ethyl 3-bromoimidazo[1,5-a]pyridine-1-carboxylate

  • MDL number MFCD17926422
  • Cas Number : 138891-58-4
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8203

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8203
Availability details for
Quantity
  • Availability
  • Add to Cart