COM448683196 Combi-Blocks, Inc.

2-[3-(2-Amino-4-thiazolyl)phenoxy]acetic acid

  • MDL number MFCD11564607
  • Cas Number : 1082128-37-7
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8258

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8258
Availability details for
Quantity
  • Availability
  • Add to Cart