COM448683244 Combi-Blocks, Inc.

Ethyl 4-amino-1-methyl-1h-imidazole-5-carboxylate

  • MDL number MFCD11053610
  • Cas Number : 61982-18-1
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8340
Availability details for
Quantity
  • Availability
  • Add to Cart