COM448683253 Combi-Blocks, Inc.

Ethyl 4-chloro-1-m-tolyl-1h-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD09788600
  • Cas Number : 951624-03-6
  • Molecular Weight : 315.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8355

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8355
Availability details for
Quantity
  • Availability
  • Add to Cart