COM448683289 Combi-Blocks, Inc.

1-(4-Bromophenyl)-4-methylpiperazine hydrobromide

  • MDL number MFCD12198861
  • Cas Number : 1186663-30-8
  • Molecular Weight : 336.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8407
Availability details for
Quantity
  • Availability
  • Add to Cart