COM448683294 Combi-Blocks, Inc.

Ethyl 4-cyanopyrrole-2-carboxylate

  • MDL number MFCD10696319
  • Cas Number : 944901-09-1
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8415
Availability details for
Quantity
  • Availability
  • Add to Cart