COM448683308 Combi-Blocks, Inc.

3,5-Dimethyl-1H-1,2,4-triazole

  • MDL number MFCD00656686
  • Cas Number : 7343-34-2
  • Molecular Weight : 97.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8433
Availability details for
Quantity
  • Availability
  • Add to Cart