COM448683317 Combi-Blocks, Inc.

Ethyl 4-oxo-1-phenyl-3-piperidinecarboxylate

  • MDL number MFCD18711416
  • Cas Number : 1026-47-7
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8452
Availability details for
Quantity
  • Availability
  • Add to Cart