COM448683341 Combi-Blocks, Inc.

Ethyl 5,7-difluoro-3-iodo-1h-indole-2-carboxylate

  • MDL number MFCD20261488
  • Cas Number : 1334499-90-9
  • Molecular Weight : 351.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8502
Availability details for
Quantity
  • Availability
  • Add to Cart