COM448683426 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-methoxyaniline

  • MDL number MFCD08444539
  • Cas Number : 7270-83-9
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8672

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8672
Availability details for
Quantity
  • Availability
  • Add to Cart