COM448683442 Combi-Blocks, Inc.

Ethyl 5-hydroxypyrazolo[1,5-a]pyrimidine-2-carboxylate

  • MDL number MFCD21607427
  • Cas Number : 1363405-50-8
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8705
Availability details for
Quantity
  • Availability
  • Add to Cart