COM448683477 Combi-Blocks, Inc.

Ethyl 6-fluoro-1-hydroxy-2-methyl-indole-3-carboxylate

  • MDL number MFCD11501321
  • Cas Number : 1086398-59-5
  • Molecular Weight : 237.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8765
Availability details for
Quantity
  • Availability
  • Add to Cart