COM448683495 Combi-Blocks, Inc.

Ethyl 7-chlorocinnoline-3-carboxylate

  • MDL number MFCD17926299
  • Cas Number : 104092-54-8
  • Molecular Weight : 236.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8795
Availability details for
Quantity
  • Availability
  • Add to Cart