COM448683537 Combi-Blocks, Inc.

3-Benzylsulfamoyl-benzoic acid

  • MDL number MFCD00622724
  • Cas Number : 7326-77-4
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-8857
Availability details for
Quantity
  • Availability
  • Add to Cart